L-Leucine ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Leu-OEt.HCl

4001907.0025 4001907.0100 4001907.0500
E-2070.0025BAEA 185.3 CAD
E-2070.0025BA E-2070.0100BA E-2070.0500BA
L-Leucine ethyl ester hydrochloride
L-Leucine ethyl ester hydrochloride

Sequence: H-Leu-OEt · HCl


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₈H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00034879
CAS Number: 2743-40-0
EINECS: 220-375-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR