L-Leucine-tert-butyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, H-Leu-OtBu·HCl, L-leucine 1,1-dimethylethyl ester hydrochloride, H-Leu-OtBu.HCl, TERT-BUTYL (2S)-2-AMINO-4-METHYLPENTANOATE HYDROCHLORIDE

4001167.0005 4001167.0025
E-2065.0005BAEA 211.77 CAD
E-2065.0005BA E-2065.0025BA
L-Leucine-tert-butyl ester hydrochloride
L-Leucine-tert-butyl ester hydrochloride

Sequence: H-Leu-OtBu · HCl


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₁₀H₂₁NO₂
MW: 223.74 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038906
CAS Number: 2748-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR