L(+)-Leucine

Supplier: BACHEM AMERICAS INC

4030792.0250 4030792.1000
E-2050.0250BAEA 175.66 CAD
E-2050.0250BA E-2050.1000BA
L(+)-Leucine
L(+)-Leucine

Sequence: H-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR