L-Isoleucine methyl ester hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ile-OMe·HCl, H-Ile-OMe.HCl
4000501.0025
4000501.0100
E-2045.0025BAEA
335.58
CAD
E-2045.0025BA
E-2045.0100BA
L-Isoleucine methyl ester hydrochloride
L-Isoleucine methyl ester hydrochloride
Sequence: H-Ile-OMe · HCl
Formula:
C₇H₁₅NO₂ MW: 181.66 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038911 CAS Number: 18598-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...