L(+)-Isoleucine

Supplier: BACHEM AMERICAS INC

4030756.0100 4030756.0500
E-2025.0100BAEA 276.63 CAD
E-2025.0100BA E-2025.0500BA
L(+)-Isoleucine
L(+)-Isoleucine

Sequence: H-Ile-OH CAS-No.: 73-32-5 Mol.weight: 131.18 SumFormula: C₆H₁₃NO₂ Bulk item no.: E-2025 Synonym(s): EINECS no.: 200-798-2 MDL no.: MFCD00064222


Sequence: H-Ile-OH

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR