L-Histidine methyl ester dihydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl
Sequence: H-His-OMe · 2 HCl
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
[1499-46-3] (net).
Formula:
C₇H₁₁N₃O₂ MW: 242.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012701 CAS Number: 7389-87-9 EINECS: 230-973-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...