H-His-NH₂·2HCl

Supplier: BACHEM AMERICAS INC

4003763.0001 4003763.0005
E-2015.0001BAEA 180.91 CAD
E-2015.0001BA E-2015.0005BA
H-His-NH₂·2HCl
H-His-NH₂·2HCl

Sequence: H-His-NH₂ · 2 HCl


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₆H₁₂Cl₂N₄O
Storage Temperature: Ambient
MDL Number: MFCD00058559
CAS Number: 71666-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR