Benzyl-2-aminoacetate
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Gly-OBzl·HCl, Glycine Benzyl Ester Hydrochloride
4010087.0005
4010087.0025
4010087.0100
E-1970.0005BAEA
180.91
CAD
E-1970.0005BA
E-1970.0025BA
E-1970.0100BA
Benzyl-2-aminoacetate
Benzyl-2-aminoacetate
Sequence: H-Gly-OBzl · HCl
Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
[2462-31-9] (hydrochloride)
Formula:
C₉H₁₁NO₂ MW: 165.192 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00035442 CAS Number: 1738-68-7 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...