H-Gln-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: L-Glutamine tert-butyl ester hydrochloride, H-Gln-OtBu.HCl, H-Gln-OtBu·HCl

4003777.0001 4003777.0005
E-1920.0001BAEA 206.44 CAD
E-1920.0001BA E-1920.0005BA
H-Gln-OtBu·HCl
H-Gln-OtBu·HCl

Sequence: H-Gln-OtBu · HCl


For isoglutamine derivatives please see the family "Glutamic Acid".


Substrate for glutaminyl cyclase (QC). For papaya QC Km 223 µm, kcat 49.4 s⁻¹, and kcat/Km 222 µm⁻¹s⁻¹ were determined.

Formula: C₉H₁₉ClN₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00039081
CAS Number: 39741-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR