L-Glutamic acid 5-methyl ester

Supplier: BACHEM AMERICAS INC

4002301.0005 4002301.0025 4002301.0100
E-1910.0005BAEA 180.91 CAD
E-1910.0005BA E-1910.0025BA E-1910.0100BA
L-Glutamic acid 5-methyl ester
L-Glutamic acid 5-methyl ester

Sequence: H-Glu(OMe)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₆H₁₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00002632
CAS Number: 1499-55-4
EINECS: 216-110-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR