H-Glu(OtBu)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

4000156.0001 4000156.0005 4000156.0025
E-1900.0001BAEA 185.46 CAD
E-1900.0001BA E-1900.0005BA E-1900.0025BA
H-Glu(OtBu)-OtBu·HCl
H-Glu(OtBu)-OtBu·HCl

Sequence: H-Glu(OtBu)-OtBu · HCl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₃H₂₆ClNO₄
Storage Temperature: Freezer
MDL Number: MFCD00058003
CAS Number: 32677-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR