H-Glu(OBzl)-OBzl.TosOH

Supplier: BACHEM AMERICAS INC

4000238.0050 4000238.0250
E-1895.0050BAEA 238.17 CAD
E-1895.0050BA E-1895.0250BA
H-Glu(OBzl)-OBzl.TosOH
H-Glu(OBzl)-OBzl.TosOH

Sequence: H-Glu(OBzl)-OBzl · p-tosylate


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₂₆H₂₉NO₇S
Storage Temperature: Refrigerator
MDL Number: MFCD00038980
CAS Number: 2791-84-6
EINECS: 220-522-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR