5-tert-Butyl-1-methyl-L-glutamate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe·HCl, H-Glu(OtBu)-OMe.HCl

4004027.0001 4004027.0005
E-1885.0001BAEA 284.04 CAD
E-1885.0001BA E-1885.0005BA
5-tert-Butyl-1-methyl-L-glutamate hydrochloride
5-tert-Butyl-1-methyl-L-glutamate hydrochloride

Sequence: H-Glu(OtBu)-OMe · HCl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₀H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00153438
CAS Number: 6234-01-1
EINECS: 228-348-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR