5-tert-Butyl-L-glutamate hydrate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Glu(OtBu)-OH, L-Glutamic Acid 5-tert-Butyl Ester

4002937.0001 4002937.0005 4002937.0025
E-1875.0001BAEA 180.91 CAD
E-1875.0001BA E-1875.0005BA E-1875.0025BA
5-tert-Butyl-L-glutamate hydrate
5-tert-Butyl-L-glutamate hydrate

Sequence: H-Glu(OtBu)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038580
CAS Number: 2419-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR