L-Glutamic acid 5-benzyl ester

Supplier: BACHEM AMERICAS INC

4000146.0005 4000146.0025 4000146.0100
E-1870.0005BAEA 180.91 CAD
E-1870.0005BA E-1870.0025BA E-1870.0100BA
L-Glutamic acid 5-benzyl ester
L-Glutamic acid 5-benzyl ester

Sequence: H-Glu(OBzl)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₂H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00002633
CAS Number: 1676-73-9
EINECS: 216-826-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR