1-Benzyl-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: L-Glutamic Acid 1-Benzyl Ester, H-Glu-OBzl, (S)-4-Amino-5-(benzyloxy)-5-oxopentanoic acid

4000337.0005 4000337.0025
E-1865.0005BAEA 439.41 CAD
E-1865.0005BA E-1865.0025BA
1-Benzyl-L-glutamate
1-Benzyl-L-glutamate

Sequence: H-Glu-OBzl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037312
CAS Number: 13030-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR