L(+)-Glutamic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

4030627.0250 4030627.0500
E-1860.0250BAEA 232.96 CAD
E-1860.0250BA E-1860.0500BA
L(+)-Glutamic acid
L(+)-Glutamic acid

Sequence: H-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR