N-Formyl-L-methionine

Supplier: BACHEM AMERICAS INC

4001407.0005 4001407.0025
E-1850.0005BAEA 206.51 CAD
E-1850.0005BA E-1850.0025BA
N-Formyl-L-methionine
N-Formyl-L-methionine

Sequence: For-Met-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.

Formula: C₆H₁₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00021033
CAS Number: 4289-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR