H-Lys(Me)₂-OH · HCl

Supplier: BACHEM AMERICAS INC

4004407.0250 4004407.1000 4004407.5000
E-1810.0250BAEA 301.72 CAD
E-1810.0250BA E-1810.1000BA E-1810.5000BA
H-Lys(Me)₂-OH · HCl
H-Lys(Me)₂-OH · HCl

Sequence: H-Lys(Me)₂-OH · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₈H₁₈N₂O₂·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00038975
CAS Number: 2259-86-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR