AC-Lys(AC)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Nalpha,epsilon-Bis-acetyl-L-lysine
4025494.0001
4025494.0005
E-1775.0001BAEA
211.67
CAD
E-1775.0001BA
E-1775.0005BA
AC-Lys(AC)-OH
AC-Lys(AC)-OH
Sequence: Ac-Lys(Ac)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₁₀H₁₈N₂O₄ MW: 230.264 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00236760 CAS Number: 499-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...