H-Lys(Z)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys(Z)-OtBu·HCl, (S)-tert-Butyl 2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

4000312.0001 4000312.0005 4000312.0025
E-1710.0001BAEA 180.91 CAD
E-1710.0001BA E-1710.0005BA E-1710.0025BA
H-Lys(Z)-OtBu·HCl
H-Lys(Z)-OtBu·HCl

Sequence: H-Lys(Z)-OtBu · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₈H₂₉ClN₂O₄
MW: 372.89 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038898
CAS Number: 5978-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR