Nε-carbobenzoxy-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH

4007216.0025 4007216.0100
E-1702.0025BAEA 180.91 CAD
E-1702.0025BA E-1702.0100BA
Nε-carbobenzoxy-L-lysine
Nepsilon-carbobenzoxy-L-lysine

Sequence: H-Lys(Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₄H₂₀N₂O₄
MW: 280.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002638
CAS Number: 1155-64-2
EINECS: 214-585-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR