H-Tyr(tBu)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Tyr(tBu)-OtBu.HCl, H-Tyr(tBu)-OtBu·HCl

4010073.0001 4010073.0005 4010073.0025
E-1690.0001BAEA 180.91 CAD
E-1690.0001BA E-1690.0005BA E-1690.0025BA
H-Tyr(tBu)-OtBu·HCl
H-Tyr(tBu)-OtBu·HCl

Sequence: H-Tyr(tBu)-OtBu · HCl


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₇H₂₈ClNO₃
Storage Temperature: Freezer
MDL Number: MFCD00058002
CAS Number: 17083-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR