H-Thr(tBu)-OMe·HCl

Supplier: BACHEM AMERICAS INC

4000336.0005 4000336.0025
E-1680.0005BAEA 284.04 CAD
E-1680.0005BA E-1680.0025BA
H-Thr(tBu)-OMe·HCl
H-Thr(tBu)-OMe·HCl

Sequence: H-Thr(tBu)-OMe · HCl


Bachem also offers phosphothreonine derivatives.

Formula: C₉H₂₀ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00077111
CAS Number: 71989-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR