O-tert-Butyl-L-serine methyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Ser(tBu)-OMe·HCl, H-Ser(tBu)-OMe.HCl

4002681.0001 4002681.0005 4002681.0025
E-1665.0001BAEA 180.91 CAD
E-1665.0001BA E-1665.0005BA E-1665.0025BA
O-tert-Butyl-L-serine methyl ester hydrochloride
O-tert-Butyl-L-serine methyl ester hydrochloride

Sequence: H-Ser(tBu)-OMe · HCl


Bachem also offers derivatives of phosphoserine and sulfoserine.


Educt for the synthesis of heterocyclic serine dipeptide analogs.

Formula: C₈H₁₇NO₃
MW: 211.69 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077108
CAS Number: 17114-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR