H-Ser(tBu)-OtBu
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-tert-Butyl 2-amino-3-(tert-butoxy)propanoate
4002610.0001
4002610.0005
4002610.0025
E-1660.0001BAEA
180.91
CAD
E-1660.0001BA
E-1660.0005BA
E-1660.0025BA
H-Ser(tBu)-OtBu
H-Ser(tBu)-OtBu
Sequence: H-Ser(tBu)-OtBu
Bachem also offers derivatives of phosphoserine and sulfoserine.
Formula:
C₁₁H₂₃NO₃ MW: 217.309 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00080900 CAS Number: 48067-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...