O-tert-Butyl-L-serine
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ser(tBu)-OH
4001695.0001
4001695.0005
4001695.0025
E-1655.0001BAEA
185.3
CAD
E-1655.0001BA
E-1655.0005BA
E-1655.0025BA
O-tert-Butyl-L-serine
O-tert-Butyl-L-serine
Sequence: H-Ser(tBu)-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00066088 CAS Number: 18822-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...