H-Orn(boc)-OH

Supplier: BACHEM AMERICAS INC

4004448.0005 4004448.0025
E-1625.0005BAEA 317.74 CAD
E-1625.0005BA E-1625.0025BA
H-Orn(boc)-OH
H-Orn(boc)-OH

Sequence: H-Orn(Boc)-OH

Formula: C₁₀H₂₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00037219
CAS Number: 13650-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR