H-Lys(boc)-OMe·HCl

Supplier: BACHEM AMERICAS INC

4003757.0001 4003757.0005 4003757.0025
E-1620.0001BAEA 180.91 CAD
E-1620.0001BA E-1620.0005BA E-1620.0025BA
H-Lys(boc)-OMe·HCl
H-Lys(boc)-OMe·HCl

Sequence: H-Lys(Boc)-OMe · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₂H₂₅ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00076959
CAS Number: 2389-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR