H-Lys(boc)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, H-Lys(Boc)-OtBu.HCl, H-Lys(Boc)-OtBu·HCl

4003765.0001 4003765.0005 4003765.0025
E-1615.0001BAEA 180.91 CAD
E-1615.0001BA E-1615.0005BA E-1615.0025BA
H-Lys(boc)-OtBu·HCl
H-Lys(boc)-OtBu·HCl

Sequence: H-Lys(Boc)-OtBu · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038899
CAS Number: 13288-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR