Nε-(tert-butoxycarbonyl)-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: Nε-Boc-L-lysine, H-Lys(Boc)-OH

4000211.0005 4000211.0025
E-1610.0005BAEA 241.04 CAD
E-1610.0005BA E-1610.0025BA
Nε-(tert-butoxycarbonyl)-L-lysine
Nepsilon-(tert-butoxycarbonyl)-L-lysine

Sequence: H-Lys(Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037221
CAS Number: 2418-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR