H-His(Bzl)-OMe · 2 HCl mixture of isomers
Supplier: BACHEM AMERICAS INC
4010041.0001 4010041.0005
E-1560.0001BAEA 180.91 CAD
E-1560.0001BA E-1560.0005BA
H-His(Bzl)-OMe · 2 HCl mixture of isomers
H-His(Bzl)-OMe · 2 HCl
Sequence: H-His(Bzl)-OMe · 2 HCl
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Mix of τ-Bzl (mainly formed) and π-Bzl isomer. CAS Number (τ-isomer): 93983-56-3.