Dimethyl-L-aspartate hydrochloride

Supplier: BACHEM AMERICAS INC

4003614.0005 4003614.0025
E-1440.0005BAEA 206.51 CAD
E-1440.0005BA E-1440.0025BA
Dimethyl-L-aspartate hydrochloride
Dimethyl-L-aspartate hydrochloride

Sequence: H-Asp(OMe)-OMe · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₆H₁₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00038878
CAS Number: 32213-95-9
EINECS: 250-957-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR