Di-tert-butyl L-aspartate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Asp(OtBu)-OtBu.HCl, H-Asp(OtBu)-OtBu·HCl, L-Aspartic Acid Di-tert-butyl Ester Hydrochloride, (S)-Di-tert-butyl 2-aminosuccinate hydrochloride

4000171.0005 4000171.0025
E-1435.0005BAEA 361.41 CAD
E-1435.0005BA E-1435.0025BA
Di-tert-butyl L-aspartate hydrochloride
Di-tert-butyl L-aspartate hydrochloride

Sequence: H-Asp(OtBu)-OtBu · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₂H₂₃NO₄
MW: 281.78 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00034851
CAS Number: 1791-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR