1,4-Dibenzyl-L-aspartate-p-toluenesulfonate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Asp(OBzl)-Obzl.TosOH, L-Aspartic Acid Dibenzyl Ester p-Toluenesulphonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulfonate, L-Aspartic acid dibenzyl ester p-toluenesulphonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulphonate, H-Asp(OBzl)-OBzl.Tos-OH, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulphonate, H-Asp(OBzl)-OBzl·TosOH, L-Aspartic acid dibenzyl ester p-toluenesulphonate, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulfonate, L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulfonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulfonate

4001090.0025 4001090.0100
E-1430.0025BAEA 180.91 CAD
E-1430.0025BA E-1430.0100BA
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate

Sequence: H-Asp(OBzl)-OBzl · p-tosylate


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₈H₁₉NO₄
MW: 485.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065188
CAS Number: 2886-33-1
EINECS: 220-746-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR