L-Asparaginamide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Asn-NH₂·HCl

4000593.0500 4000593.1000
E-1425.0500BAEA 180.91 CAD
E-1425.0500BA E-1425.1000BA
L-Asparaginamide hydrochloride
L-Asparaginamide hydrochloride

Sequence: H-Asn-NH₂ · HCl


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₄H₁₀ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00077102
CAS Number: 57471-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR