H-Asp(OtBu)-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, H-Asp(OtBu)-OMe.HCl

4004029.0001 4004029.0005
E-1420.0001BAEA 413.58 CAD
E-1420.0001BA E-1420.0005BA
H-Asp(OtBu)-OMe·HCl
H-Asp(OtBu)-OMe·HCl

Sequence: H-Asp(OtBu)-OMe · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₉H₁₈ClNO₄
MW: 239.7 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153427
CAS Number: 2673-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR