(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 4-tert-Butyl L-Aspartate, H-Asp(OtBu)-OH, L-Aspartic acid 4-tert-butyl ester, L-Aspartic acid 4-tert-Butyl ester

4002930.0005 4002930.0025
E-1410.0005BAEA 529.57 CAD
E-1410.0005BA E-1410.0025BA
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid
(S)-2-Amino-4-(tert-butoxy)-4-oxobutanoic acid

Sequence: H-Asp(OtBu)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038577
CAS Number: 3057-74-7
EINECS: 221-292-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR