4-Benzyl-L-aspartate

Supplier: BACHEM AMERICAS INC

Synonyms: L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester

4000257.0025 4000257.0100
E-1405.0025BAEA 490.51 CAD
E-1405.0025BA E-1405.0100BA
4-Benzyl-L-aspartate
4-Benzyl-L-aspartate

Sequence: H-Asp(OBzl)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037208
CAS Number: 2177-63-1
EINECS: 218-541-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR