L(+)-Aspartic acid

Supplier: BACHEM AMERICAS INC

4030334.0250 4030334.1000
E-1395.0250BAEA 180.07 CAD
E-1395.0250BA E-1395.1000BA
L(+)-Aspartic acid
L(+)-Aspartic acid

Sequence: H-Asp-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₄H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR