H-Asn-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Asn-OtBu·HCl, L-Asparagine tert-butyl ester hydrochloride

4000340.0001 4000340.0005
E-1390.0001BAEA 206.51 CAD
E-1390.0001BA E-1390.0005BA
H-Asn-OtBu·HCl
H-Asn-OtBu·HCl

Sequence: H-Asn-OtBu · HCl


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₈H₁₇ClN₂O₃
MW: 224.685 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039079
CAS Number: 63094-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR