H-Asn-OtBu

Supplier: BACHEM AMERICAS INC

4030375.0001 4030375.0005
E-1385.0001BAEA 232.35 CAD
E-1385.0001BA E-1385.0005BA
H-Asn-OtBu
H-Asn-OtBu

Sequence: H-Asn-OtBu


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₈H₁₆N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00190755
CAS Number: 25456-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR