L-Arginine methyl ester dihydrochloride

Supplier: BACHEM AMERICAS INC

4002664.0025 4002664.0100
E-1375.0025BAEA 206.51 CAD
E-1375.0025BA E-1375.0100BA
L-Arginine methyl ester dihydrochloride
L-Arginine methyl ester dihydrochloride

Sequence: H-Arg-OMe · 2 HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₇H₁₆N₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00038948
CAS Number: 26340-89-6
EINECS: 247-622-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR