L-Argininamide dihydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Arg-NH₂·2HCl, H-Arg-NH₂·2 HCl, H-Arg-NH2.2HCl

4007034.0005 4007034.0025 4007034.0100
E-1365.0005BAEA 206.51 CAD
E-1365.0005BA E-1365.0025BA E-1365.0100BA
L-Argininamide dihydrochloride
L-Argininamide dihydrochloride

Sequence: H-Arg-NH₂ · 2 HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₆H₁₇Cl₂N₅O
MW: 246.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058286
CAS Number: 14975-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR