H-Cys(aminoethyl)-OH · HCl

Supplier: BACHEM AMERICAS INC

4001430.0001 4001430.0005
E-1355.0001BAEA 180.91 CAD
E-1355.0001BA E-1355.0005BA
H-Cys(aminoethyl)-OH · HCl
H-Cys(aminoethyl)-OH · HCl

Sequence: H-Cys(aminoethyl)-OH · HCl
Synonym(s): L-4-Thialysine · HCl#L-Thiosine · HCl


A naturally occuring cytotoxic lysine analog. Substrate of bacterial lysine-2-monooxygenase. L-Thialysine inhibits homocitrate synthase, E. coli aspartate kinase III, and the lysine aminomutases lysine-2,3-aminomutase and lysine-5,6-aminomutase.

Formula: C₅H₁₂N₂O₂S·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00036385
CAS Number: 4099-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR