L-Alanine ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

4001369.0025 4001369.0100
E-1330.0025BAEA 211.77 CAD
E-1330.0025BA E-1330.0100BA
L-Alanine ethyl ester hydrochloride
L-Alanine ethyl ester hydrochloride

Sequence: H-Ala-OEt · HCl


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₅H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00063662
CAS Number: 1115-59-9
EINECS: 214-225-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR