L-Alanine-tert-butyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

4000207.0005 4000207.0025
E-1325.0005BAEA 211.67 CAD
E-1325.0005BA E-1325.0025BA
L-Alanine-tert-butyl ester hydrochloride
L-Alanine-tert-butyl ester hydrochloride

Sequence: H-Ala-OtBu · HCl


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₇H₁₅NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00035524
CAS Number: 13404-22-3
EINECS: 236-497-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR