Ala-OBzl TosOH

Supplier: BACHEM AMERICAS INC

Synonyms: L-Alanine Benzyl Ester p-Toluenesulphonate, L-Alanine benzyl ester p-toluenesulphonate salt, salt H-Ala-OBzl.Tos-OH, H-Ala-OBzl·p-tosylate, L-Alanine benzyl ester p-toluenesulfonate salt, H-Ala-OBzl.TosOH, L-Alanine Benzyl Ester p-Toluenesulfonate, H-Ala-OBzl·Tos-OH

4001352.0025 4001352.0100
E-1315.0025BAEA 309.81 CAD
E-1315.0025BA E-1315.0100BA
Ala-OBzl TosOH
Ala-OBzl TosOH

Sequence: H-Ala-OBzl · p-tosylate


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₁₇H₂₁NO₅S
MW: 351.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00066143
CAS Number: 42854-62-6
EINECS: 255-969-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR