L-Alanine benzyl ester hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ala-OBzl.HCl, H-Ala-OBzl·HCl
4001744.0005
4001744.0025
4001744.0100
E-1310.0005BAEA
185.3
CAD
E-1310.0005BA
E-1310.0025BA
E-1310.0100BA
L-Alanine benzyl ester hydrochloride
L-Alanine benzyl ester hydrochloride
Sequence: H-Ala-OBzl · HCl
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
Formula:
C₁₀H₁₃NO₂ Storage Temperature: Freezer |
MDL Number:
MFCD00054340 CAS Number: 5557-83-5 EINECS: 226-920-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...