L-Alanine

Supplier: BACHEM AMERICAS INC

4030174.0100 4030174.0250
E-1285.0100BAEA 175.66 CAD
E-1285.0100BA E-1285.0250BA
L-Alanine
L-Alanine

Sequence: H-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₃H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR